Triiodomethane ≥99%

Warning

109452-5G 109452-100G 109452-500G
220033-845EA 43.62 USD
220033-845 220033-843 220033-844
Triiodomethane ≥99%
Triiodomethane
Formula: CHI₃
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 123 °C
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Merck Index: 13,05055

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR