Sodium benzoate ≥99%, ReagentPlus®

Synonyms: Benzoic acid sodium salt

109169-3KG 109169-1KG 109169-25G
220033-590EA 94.62 USD
220033-590 220033-588 220033-589
Sodium benzoate ≥99%, ReagentPlus®
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410 - 430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR