Isopropenyl acetate ≥99%, Sigma-Aldrich®

10914-25KG 10914-1KG
220033-582EA 1498.46 USD
220033-582 220033-581
Isopropenyl acetate ≥99%, Sigma-Aldrich®
Isopropenyl acetate

Envase: produced by Wacker Chemie AG

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.9188 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008709
CAS Number: 108-22-5
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR