Chloroacetaldehyde dimethyl acetal ≥99%, Sigma-Aldrich®

Synonyms: 1-Chloro-2,2-dimethoxyethane

10906-25KG 10906-210KG 10906-1KG
220033-572EA 2261.54 USD
220033-572 220033-815 220033-814
Chloroacetaldehyde dimethyl acetal ≥99%, Sigma-Aldrich®
Chloroacetaldehyde dimethyl acetal

Envase: produced by Wacker Chemie AG

Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 128 °C (1013 hPa)
Melting Pt: < –73 °C
Density: 1.098 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000948
CAS Number: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR