Benzyl acetoacetate ≥96%, Sigma-Aldrich®

10898-1KG 10898-25KG
220033-206EA 326.15 USD
220033-206 220033-207
Benzyl acetoacetate ≥96%, Sigma-Aldrich®
Benzyl acetoacetate

Envase: produced by Wacker Chemie AG

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00008785
CAS Number: 5396-89-4
EINECS: 226-416-4

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR