6-Methylquinoline ≥98%

108928-100G 108928-25G
220033-802EA 186.31 USD
220033-802 220033-803
6-Methylquinoline ≥98%
6-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 258.6 °C (1013 hPa)
Melting Pt: –22.2 °C
Density: 1.0815 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006804
CAS Number: 91-62-3
EINECS: 202-084-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR