4-Nitro-o-phenylenediamine ≥98%

Warning

Synonyms: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

108898-100G 108898-5G
220033-798EA 105 USD
220033-798 220033-799
4-Nitro-o-phenylenediamine ≥98%
4-Nitro-o-phenylenediamine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 197…202 °C
MDL Number: MFCD00007724
CAS Number: 99-56-9
EINECS: 202-766-3
Merck Index: 12,06720

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR