4-Chloro-o-phenylenediamine ≥97%

Synonyms: 1-Chloro-3,4-diaminobenzene, 4-Chlorobenzene-1,2-diamine, 4-Chloro-1,2-phenylenediamine, 4-Chloro-1,2-diaminobenzene

108871-5G 108871-25G
220033-797EA 42.89 USD
220033-797 220033-796
4-Chloro-o-phenylenediamine ≥97%
4-Chloro-o-phenylenediamine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 89…90 °C
MDL Number: MFCD00011691
CAS Number: 95-83-0
EINECS: 202-456-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR