2-Aminobenzothiazole

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

108812-100G 108812-5G
220033-789EA 81.42 USD
220033-789 220033-790
2-Aminobenzothiazole
2-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 190…195 °C (0.05 mmHg)
Melting Pt: 127…130 °C
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR