2-Methyl-3-pentanone ≥97%

Synonyms: Ethyl Isopropyl ketone

108707-25G 108707-5G
220033-774EA 198.15 USD
220033-774 220033-775
2-Methyl-3-pentanone ≥97%
2-Methyl-3-pentanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: -69 °C
Density: 0.8845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009314
CAS Number: 565-69-5
EINECS: 209-288-4
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR