4-Bromobenzoic acid ≥98%

Warning

Synonyms: p-Bromobenzoic Acid

108510-25G 108510-100G
220033-763EA 54.71 USD
220033-763 220033-762
4-Bromobenzoic acid ≥98%
4-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 252…256 °C
Density: 1.894 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 13,01392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR