2,5-Dibromothiophene

Warning

108472-100G 108472-25G
220033-755EA 146.46 USD
220033-755 220033-756
2,5-Dibromothiophene
2,5-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: –6 °C
Density: 2.1531 g/cm³ (13.8 °C)
MDL Number: MFCD00005420
CAS Number: 3141-27-3
EINECS: 221-547-3
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR