Trimethyl orthoformate ≥99%

108456-20L 108456-500ML
220033-750EA 1260 USD
220033-750 220033-751
Trimethyl orthoformate ≥99%
Trimethyl orthoformate
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: -53 °C
Density: 0.9691 g/cm³ (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II
Merck Index: 12,07012

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR