Benzenesulfonamide ≥98%

Warning

108146-5G 108146-100G 108146-500G
220033-291EA 31.88 USD
220033-291 220033-289 220033-290
Benzenesulfonamide ≥98%
Benzenesulphonamide
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 149 - 152 °C
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR