Benzenesulfonyl chloride ≥99%

Danger

Synonyms: Benzene sulphochloride

108138-1KG 108138-5G 108138-100G 108138-500G
220033-717EA 128.15 USD
220033-717 220033-719 220033-716 220033-718
Benzenesulfonyl chloride ≥99%
Benzenesulphonyl chloride
Formula: C₆H₅ClO₂S
MW: 176.62 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.3842 g/cm³ (20 °C)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 13,01072

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR