2,5-Diiodobenzoic acid ≥97%

107905-5G 107905-1G
220033-235EA 376.92 USD
220033-235 220033-234
2,5-Diiodobenzoic acid ≥97%
2,5-Diiodobenzoic acid
Formula: C₇H₄I₂O₂
MW: 373.92 g/mol
MDL Number: MFCD00045801
CAS Number: 14192-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR