α,α,α-Trifluoroacetophenone ≥99%

Danger

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

107840-100G 107840-25G 107840-5G
220033-227EA 534.15 USD
220033-227 220033-228 220033-229
α,α,α-Trifluoroacetophenone ≥99%
α,α,α-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.2799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69025716b957e6a38ed2ae6031ab9b0f10246bd1&vid=60781a8070ecc9020bf7794534c2b3fe9acf3ebf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR