Antimony (III) sulfate

Warning

Synonyms: Diantimony tris(sulphate)

10783-10G 10783-50G
220033-655EA 167.69 USD
220033-655 220033-656
Antimony (III) sulfate
Antimony(III) sulphate
Formula: O₁₂S₃Sb₂
MW: 531.71 g/mol
Density: 3.6246 g/cm³ (20 °C)
CAS Number: 7446-32-4
EINECS: 231-207-6
UN: 3077
ADR: 9,III
Merck Index: 12,00745

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR