α-Bromo-2-nitrotoluene

Danger

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

107794-25G 107794-5G
220033-651EA 136.77 USD
220033-651 220033-652
α-Bromo-2-nitrotoluene
α-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR