4-(Dimethylamino)pyridine, ReagentPlus®

Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

107700-25G 107700-100G 107700-5G
220033-639EA 42.29 USD
220033-639 220033-638 220033-640
4-(Dimethylamino)pyridine, ReagentPlus®
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 109…111 °C
Flash Pt: 124 °C
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR