Sodium formate ≥97%, Reagent Grade

Synonyms: Formic acid sodium salt

107603-1KG 107603-4KG
220033-565EA 33.42 USD
220033-565 220033-566
Sodium formate ≥97%, Reagent Grade
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 257.31 °C
Density: 1.919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR