Pyruvic acid ≥98%

Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

107360-100G 107360-25G
220032-147EA 88.74 USD
220032-147 220032-148
Pyruvic acid ≥98%
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.58 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR