(±)-3-Chloro-1,2-propanediol ≥98%

Synonyms: (±)-3-Chloro-1,2-dihydroxy propane

107271-500ML 107271-25ML
220033-561EA 80.77 USD
220033-561 220033-560
(±)-3-Chloro-1,2-propanediol ≥98%
(±)-3-Chloro-1,2-propanediol
Formula: C₃H₇ClO₂
MW: 110.54 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: -40 °C
Density: 1.32 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00004712
CAS Number: 96-24-2
EINECS: 202-492-4
UN: 2689
ADR: 6.1,III
Merck Index: 13,02164

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR