3-Hydroxy-2-nitropyridine

Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol

107255-25G 107255-100G
220033-557EA 53.42 USD
220033-557 220033-556
3-Hydroxy-2-nitropyridine
3-Hydroxy-2-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00006258
CAS Number: 15128-82-2
EINECS: 239-191-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR