Diethyl sulfide ≥98%

107247-5ML 107247-100ML
220033-555EA 47.17 USD
220033-555 220033-554
Diethyl sulfide ≥98%
Diethyl sulphide
Formula: C₄H₁₀S
MW: 90.19 g/mol
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –104 °C
Density: 0.837 g/cm³ (20 °C)
MDL Number: MFCD00009270
CAS Number: 352-93-2
EINECS: 206-526-9
UN: 2375
ADR: 3,II
Merck Index: 13,03885

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR