Adamantane-1-carboxylic acid

106399-100G 106399-25G
220033-285EA 118.46 USD
220033-285 220033-286
Adamantane-1-carboxylic acid
Adamantane-1-carboxylic acid
Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Melting Pt: 172…175 °C
Density: 1.221 g/cm³ (20 °C)
MDL Number: MFCD00074720
CAS Number: 828-51-3
EINECS: 212-584-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR