Trifluoroacetic anhydride ≥99%, ReagentPlus®

Danger

Synonyms: Trifluoracetic anhydride, TFAA

106232-500G 106232-3KG 106232-100G 106232-25KG 106232-25G
220033-495EA 121.85 USD
220033-495 220033-494 220033-491 220033-493 220033-492
Trifluoroacetic anhydride ≥99%, ReagentPlus®
Trifluoroacetic anhydride
Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39.4 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.487 g/cm³ (20 °C)
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR