3-Bromothiophene

Synonyms: 3-Thienyl bromide

106224-100G 106224-5G 106224-25G
220033-488EA 172.31 USD
220033-488 220033-490 220033-489
3-Bromothiophene
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR