2,4-Dichlorophenol ≥99%

Danger

Synonyms: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene

105953-5G 105953-250G 105953-100G
220030-814EA 35.32 USD
220030-814 220030-813 220030-812
2,4-Dichlorophenol ≥99%
2,4-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 40…41 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002169
CAS Number: 120-83-2
EINECS: 204-429-6
UN: 2928
ADR: 6.1,II
Merck Index: 13,03098

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR