α,α'-Dichloro-p-xylene ≥98%

Synonyms: 1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride

105740-25G 105740-100G
220033-480EA 38.12 USD
220033-480 220033-479
α,α'-Dichloro-p-xylene ≥98%
α,α'-Dichloro-p-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 100 °C
Density: 1.432 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000920
CAS Number: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR