2-Amino-2',5-dichlorobenzophenone ≥99%

Synonyms: (2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone

105155-50G 105155-10G
220033-457EA 191.69 USD
220033-457 220033-456
2-Amino-2',5-dichlorobenzophenone ≥99%
2-Amino-2',5-dichlorobenzophenone
Formula: C₁₃H₉Cl₂NO
MW: 266.13 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 88 - 91 °C
MDL Number: MFCD00007840
CAS Number: 2958-36-3
EINECS: 220-985-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR