1,6-Bis(dimethylamino)hexane ≥99%

Synonyms: N,N,N',N'-Tetramethylhexamethylenediamine, N,N,N',N'-Tetramethyl-1,6-diaminohexane, N1,N1,N6,N6-Tetramethylhexane-1,6-diamine, N,N,N',N'-Tetramethyl-1,6-hexanediamine

105139-1L 105139-100ML
220033-455EA 1064.46 USD
220033-455 220033-454
1,6-Bis(dimethylamino)hexane ≥99%
1,6-Bis(dimethylamino)hexane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: -46 °C
Density: 0.8064 g/cm³ (15 °C)
MDL Number: MFCD00008339
CAS Number: 111-18-2
EINECS: 203-842-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR