Octadecyltrichlorosilane

Danger

Synonyms: Trichloro(stearyl)silane, Trichlorooctadecylsilane, Octadecyltrichlorosilane, ODTS, Trichloro(octadecyl)silane

104817-100G 104817-25G 104817-500G
220033-435EA 517.85 USD
220033-435 220033-436 220033-437
Octadecyltrichlorosilane
Octadecyltrichlorosilane
Formula: C₁₈H₃₇Cl₃Si
MW: 387.94 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 22 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00000484
CAS Number: 112-04-9
EINECS: 203-930-7
UN: 1800
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR