4-Nitrophenol for spectrophotometry

Warning

1048-25G 1048-5G
220033-433EA 219.23 USD
220033-433 220033-434
4-Nitrophenol for spectrophotometry
4-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
EINECS: 202-811-7
UN: 1663
ADR: 6.1,III
Merck Index: 13,06654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR