Tetrafluoro-p-benzoquinone

Synonyms: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

104353-1G 104353-5G
220033-421EA 43.08 USD
220033-421 220033-422
Tetrafluoro-p-benzoquinone
Tetrafluoro-p-benzoquinone
Formula: C₆F₄O₂
MW: 180.06 g/mol
Boiling Pt: 133 °C (1013 hPa)
MDL Number: MFCD00001592
CAS Number: 527-21-9
EINECS: 208-411-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR