Ethyl toluene-4-sulfonate ≥98%

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

104256-1KG 104256-50G 104256-250G
220033-416EA 246.62 USD
220033-416 220033-418 220033-417
Ethyl toluene-4-sulfonate ≥98%
Ethyl toluene-4-sulphonate
Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009100
CAS Number: 80-40-0
EINECS: 201-276-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,03889

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR