Pentafluorophenol ≥99%

Warning

Synonyms: 2,3,4,5,6-Pentafluorophenol

103799-100G 103799-10G 103799-5G
220033-383EA 326.31 USD
220033-383 220033-384 220033-385
Pentafluorophenol ≥99%
Pentafluorophenol
Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: 34…36 °C
Density: 1.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR