Pentafluorobenzenesulfonyl chloride ≥99%

Synonyms: 2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride

103764-25G 103764-5G
220032-144EA 170.15 USD
220032-144 220033-379
Pentafluorobenzenesulfonyl chloride ≥99%
Pentafluorobenzenesulphonyl chloride
Formula: C₆ClF₅O₂S
MW: 266.58 g/mol
Boiling Pt: 210 °C (1013 hPa)
Density: 1.79 g/cm³ (20 °C)
MDL Number: MFCD00007427
CAS Number: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR