(3-Nitrophenyl)acetic acid ≥99%

Warning

103608-1G 103608-10G 103608-5G
220032-041EA 2288 USD
220032-041 220032-040 220032-135
(3-Nitrophenyl)acetic acid ≥99%
(3-Nitrophenyl)acetic acid
Formula: (C₆H₄(NO₂))CH₂COOH
MW: 181.15 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00007278
CAS Number: 1877-73-2
EINECS: 217-512-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR