4-Nitroanisole ≥97%

103543-25G 103543-5G
220032-036EA 59.45 USD
220032-036 220032-037
4-Nitroanisole ≥97%
4-Nitroanisole
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 9.4 °C
Density: 1.254 g/cm³ (20 °C)
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 12,06682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR