6-Methoxy-m-toluidine ≥99%

Danger

Synonyms: 3-Amino-4-methoxytoluene, 2-Amino-4-methylanisole, 1-Amino-2-methoxy-5-methylbenzene, 5-Methyl-o-anisidine, p-Cresidine, 2-Methoxy-5-methylaniline

103284-500G 103284-100G
220033-373EA 468.46 USD
220033-373 220033-372
6-Methoxy-m-toluidine ≥99%
6-Methoxy-m-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 55.5 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00007815
CAS Number: 120-71-8
EINECS: 204-419-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR