Pentafluoroiodobenzene ≥99%

Warning

Synonyms: Iodopentafluorobenzene

103136-5G 103136-25G
220033-360EA 36.08 USD
220033-360 220033-359
Pentafluoroiodobenzene ≥99%
Pentafluoroiodobenzene
Formula: C₆F₅I
MW: 293.96 g/mol
Boiling Pt: 161…163 °C (1013 hPa)
Melting Pt: –29 °C
Density: 2.2055 g/cm³ (25 °C)
MDL Number: MFCD00001032
CAS Number: 827-15-6
EINECS: 212-565-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR