Bis(4-hydroxyphenyl) sulfone ≥98%

Synonyms: 4,4'-Sulphonyldiphenol, 4,4'-Dihydroxy diphenyl sulphone, 4-Hydroxyphenyl sulphone, 4,4'-sulphonyldiphenol

103039-100G 103039-500G
220033-353EA 61.58 USD
220033-353 220033-354
Bis(4-hydroxyphenyl) sulfone ≥98%
Bis(4-hydroxyphenyl)sulphone
Formula: C₁₂H₁₀O₄S
MW: 250.28 g/mol
MDL Number: MFCD00002350
CAS Number: 80-09-1
EINECS: 201-250-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR