3-Hexanone ≥98%

Danger

103020-10G 103020-100G
220033-352EA 242.31 USD
220033-352 220033-351
3-Hexanone ≥98%
3-Hexanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: -55.4 °C
Density: 0.8216 g/cm³ (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009402
CAS Number: 589-38-8
EINECS: 209-645-4
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR