3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥97%

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

103012-1G 103012-5G
220033-348EA 42.97 USD
220033-348 220033-349
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥97%
3-Hydroxy-4-methoxycinnamic acid
Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: 180 - 182 °C
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR