2-Hexanone ≥98%, Reagent Grade

Danger

Synonyms: Methyl butyl ketone, Butyl methyl ketone

103004-50G 103004-10G 103004-250G
220033-345EA 49.54 USD
220033-345 220033-343 220033-344
2-Hexanone ≥98%, Reagent Grade
2-Hexanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: -56.9 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009482
CAS Number: 591-78-6
EINECS: 209-731-1
UN: 1224
ADR: 3,III
Merck Index: 13,06060

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR