2-Fluorobiphenyl ≥96%

Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1

102741-10G 102741-1G
220033-337EA 137.85 USD
220033-337 220033-338
2-Fluorobiphenyl ≥96%
2-Fluorobiphenyl
Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 73 - 74 °C
Density: 1.2452 g/cm³ (25 °C)
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR