2,4-Diaminotoluene ≥98%

Danger

Synonyms: 4-Methyl-1,3-Phenylenediamine, 1,3-Diamino-4-methylbenzene, Toluene-2,4-diamine

101915-250G 101915-50G
220032-587EA 295.54 USD
220032-587 220032-588
2,4-Diaminotoluene ≥98%
2,4-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 99 °C
Density: 1.045 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007804
CAS Number: 95-80-7
EINECS: 202-453-1
UN: 1709
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR