Cyclohexanemethylamine ≥98%

Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

101842-5G 101842-25G
220033-198EA 62.68 USD
220033-198 220033-197
Cyclohexanemethylamine ≥98%
Cyclohexanemethylamine
Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 161…163 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.871 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001520
CAS Number: 3218-02-8
EINECS: 221-741-8
UN: 2735
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR