Cyclohexanecarboxylic acid ≥98%

Warning

Synonyms: Hexahydrobenzoic acid

101834-5G 101834-500G 101834-100G
220033-196EA 34.25 USD
220033-196 220033-195 220032-112
Cyclohexanecarboxylic acid ≥98%
Cyclohexanecarboxylic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 31 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5
EINECS: 202-711-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR